siRNA Details
| Gene Symbol: Sppl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029012.2 | NP_083288.2 | 11 | 1853 | ||
| GenBank mRNA | BC023131.1 | - | 7 | 1468 | ||
| BC078437.1 | - | 2 | 1900 | |||
| BC086646.1 | - | 11 | 2218 | 6817386 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sppl3| Gene Symbol: Sppl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029012.2 | NP_083288.2 | 11 | 1853 | ||
| GenBank mRNA | BC023131.1 | - | 7 | 1468 | ||
| BC078437.1 | - | 2 | 1900 | |||
| BC086646.1 | - | 11 | 2218 | 6817386 | ||