siRNA Details
| Gene Symbol: Rad54l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030730.2 | NP_109655.2 | 5, 6 | 894 | ||
| GenBank mRNA | AJ132389.1 | - | 5, 6 | 795 | ||
| AK083903.1 | - | 5, 6 | 875 | |||
| AK143506.1 | - | 4, 5 | 510 | |||
| AK155151.1 | - | 5, 6 | 851 | |||
| BC133714.1 | - | 5, 6 | 743 | 40143565 | ||
| X97574.1 | - | 3, 4 | 398 | |||

Rad54l2
Genomic Map