siRNA Details
| Gene Symbol: Tssk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032004.1 | NP_114393.1 | 1 | 506 | ||
| GenBank mRNA | AF329484.1 | - | 1 | 506 | ||
| AK076653.1 | - | 1 | 233 | |||
| BC049542.1 | - | 1 | 537 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106078.

Tssk6
Genomic Map