siRNA Details
| Gene Symbol: Ehbp1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF305087.1 | - | 12 | 4685 | ||
| AF305088.1 | - | 12 | 2159 | |||
| AF305089.1 | - | 11 | 1814 | |||
| AF305090.1 | - | 12 | 1850 | |||
| AK089917.1 | - | 11 | 1822 | |||
| AK169974.1 | - | 12 | 2410 | |||
| BC004660.1 | - | 12 | 2155 | 3497913 | ||

Ehbp1l1
Genomic Map