siRNA Details
Gene Symbol: Cd209a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133238.5 | NP_573501.1 | 6 | 608 | ||
GenBank mRNA | AF373408.1 | - | 6 | 555 | ||
AF374470.1 | - | 5 | 474 | |||
AK054139.1 | - | 6 | 572 | |||
AK171639.1 | - | 6 | 552 | |||
AY049062.1 | - | 6 | 608 | |||
BC111095.1 | - | 5 | 656 | 40040036 |
