siRNA Details
| Gene Symbol: Cd209a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133238.5 | NP_573501.1 | 7 | 942 | ||
| GenBank mRNA | AF373408.1 | - | 7 | 889 | ||
| AK054139.1 | - | 7 | 906 | |||
| AK171639.1 | - | 7 | 886 | |||
| AY049062.1 | - | 7 | 942 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd209a| Gene Symbol: Cd209a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133238.5 | NP_573501.1 | 7 | 942 | ||
| GenBank mRNA | AF373408.1 | - | 7 | 889 | ||
| AK054139.1 | - | 7 | 906 | |||
| AK171639.1 | - | 7 | 886 | |||
| AY049062.1 | - | 7 | 942 | |||