siRNA Details
Gene Symbol: Txlnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138628.3 | NP_619534.2 | 10 | 3056 | ||
GenBank mRNA | AF422245.1 | - | 10 | 2993 | ||
AK137442.1 | - | 10 | 2971 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Txlnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138628.3 | NP_619534.2 | 10 | 3056 | ||
GenBank mRNA | AF422245.1 | - | 10 | 2993 | ||
AK137442.1 | - | 10 | 2971 |