siRNA Details
| Gene Symbol: Phf21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109690.1 | NP_001103160.1 | 7 | 1001 | ||
| NM_001109691.1 | NP_001103161.1 | 10 | 1157 | |||
| NM_138755.2 | NP_620094.2 | 9 | 1160 | |||
| GenBank mRNA | AB105178.1 | - | 9 | 1066 | ||
| AK037174.1 | - | 10 | 1158 | |||
| AK129424.1 | - | 9 | 2419 | |||
| AK137085.1 | - | 6 | 968 | |||
| AY206983.1 | - | 1 | 195 | |||
| BC019181.1 | - | 9 | 1015 | 5008954 | ||

Phf21a
Genomic Map