siRNA Details
| Gene Symbol: Zmym5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253752.1 | NP_001240681.1 | 3 | 539 | ||
| NM_001253753.1 | NP_001240682.1 | 3 | 609 | |||
| NM_144842.3 | NP_659091.3 | 3 | 603 | |||
| GenBank mRNA | AK036513.1 | - | 2 | 465 | ||
| AK155333.1 | - | 3 | 521 | |||
| AK156501.1 | - | 3 | 611 | |||
| BC058644.1 | - | 2 | 400 | |||
| BU518427.1 | - | 3 | 603 | |||

Zmym5
Genomic Map