siRNA Details
| Gene Symbol: Zmym5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253752.1 | NP_001240681.1 | 6 | 1074 | ||
| NM_001253753.1 | NP_001240682.1 | 6 | 1144 | |||
| NM_144842.3 | NP_659091.3 | 6 | 1138 | |||
| GenBank mRNA | AK036513.1 | - | 5 | 1000 | ||
| AK155333.1 | - | 6 | 1056 | |||
| AK156501.1 | - | 6 | 1146 | |||
| BC027380.1 | - | 4 | 487 | |||
| BC058644.1 | - | 4 | 2014 | |||
| BE982095.1 | - | 3 | 398 | |||

Zmym5
Genomic Map