siRNA Details
| Gene Symbol: Spsb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145134.3 | NP_660116.1 | 3 | 2109 | ||
| GenBank mRNA | AF403039.2 | - | 3 | 2112 | ||
| AK142195.1 | - | 3 | 1989 | |||
| BC023083.1 | - | 2 | 1846 | 4953500 | ||
| BC055705.1 | - | 3 | 1977 | 6412566 | ||
| BC094332.1 | - | 3 | 2572 | 6817091 | ||

Spsb4
Genomic Map