siRNA Details
| Gene Symbol: Mgl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145137.2 | NP_660119.1 | 10 | 1047 | ||
| GenBank mRNA | AK085751.1 | - | 8 | 1051 | ||
| AY103461.1 | - | 10 | 984 | |||
| BC106833.1 | - | 9 | 973 | 40044847 | ||
| BC110475.2 | - | 9 | 973 | 40045765 | ||
| BC111520.1 | - | 9 | 973 | 40042306 | ||
| EU010011.1 | - | 9 | 1126 | |||

Mgl2
Genomic Map