siRNA Details
| Gene Symbol: Crebzf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145151.3 | NP_660133.2 | 1 | 1371 | ||
| GenBank mRNA | AF408398.1 | - | 1 | 1043 | ||
| BI990274.1 | - | 1 | 310 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crebzf| Gene Symbol: Crebzf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145151.3 | NP_660133.2 | 1 | 1371 | ||
| GenBank mRNA | AF408398.1 | - | 1 | 1043 | ||
| BI990274.1 | - | 1 | 310 | |||