siRNA Details
| Gene Symbol: AI661453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145489.2 | NP_663464.2 | 5 | 3867 | ||
| GenBank mRNA | BC006605.1 | - | 5 | 715 | 3157220 | |
| BC029008.1 | - | 2 | 676 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI661453| Gene Symbol: AI661453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145489.2 | NP_663464.2 | 5 | 3867 | ||
| GenBank mRNA | BC006605.1 | - | 5 | 715 | 3157220 | |
| BC029008.1 | - | 2 | 676 | |||