siRNA Details
| Gene Symbol: RASGRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128602.1 | NP_001122074.1 | 4 | 544 | ||
| NM_001306086.1 | NP_001293015.1 | 4 | 544 | |||
| NM_005739.3 | NP_005730.2 | 4 | 544 | |||
| XM_005254114.2 | XP_005254171.1 | 4 | 533 | |||
| XM_011521151.2 | XP_011519453.1 | 4 | 533 | |||
| XM_017021860.1 | XP_016877349.1 | 4 | 471 | |||
| GenBank mRNA | AF081195.1 | - | 4 | 469 | ||
| AF081197.1 | - | 3 | 323 | |||
| AF106071.1 | - | 4 | 378 | |||
| AK295097.1 | - | 3 | 408 | |||
| AK314091.1 | - | 4 | 544 | |||
| AK316067.1 | - | 4 | 472 | |||
| AY634315.2 | - | 4 | 373 | |||
| AY858556.2 | - | 4 | 373 | |||
| AY954625.4 | - | 4 | 373 | |||
| AY966005.1 | - | 4 | 373 | |||
| BC067298.1 | - | 4 | 449 | |||
| BC109296.1 | - | 4 | 416 | 40034427 | ||
| BC109297.1 | - | 4 | 416 | 40034428 | ||

RASGRP1
Genomic Map