siRNA Details
| Gene Symbol: Tmem82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145987.2 | NP_666099.2 | 6 | 1984 | ||
| GenBank mRNA | AK154809.1 | - | 4 | 1973 | ||
| AK162322.1 | - | 5 | 2404 | |||
| BC025833.1 | - | 6 | 1385 | 5125918 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem82| Gene Symbol: Tmem82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145987.2 | NP_666099.2 | 6 | 1984 | ||
| GenBank mRNA | AK154809.1 | - | 4 | 1973 | ||
| AK162322.1 | - | 5 | 2404 | |||
| BC025833.1 | - | 6 | 1385 | 5125918 | ||