siRNA Details
| Gene Symbol: Tgif2lx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153109.1 | NP_694749.1 | 1 | 604 | ||
| GenBank mRNA | AF434662.1 | - | 1 | 604 | ||
| AK132935.1 | - | 1 | 613 | |||
| BC111833.1 | - | 1 | 521 | 40041012 | ||
| Gene Symbol: Tgif2lx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142750.1 | NP_001136222.1 | 1 | 612 | ||

Tgif2lx1
Genomic Map