siRNA Details
| Gene Symbol: BC028528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153513.2 | NP_705733.2 | 4 | 234 | ||
| GenBank mRNA | AF534879.1 | - | 4 | 234 | ||
| BC028528.1 | - | 4 | 252 | 1366537 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC028528| Gene Symbol: BC028528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153513.2 | NP_705733.2 | 4 | 234 | ||
| GenBank mRNA | AF534879.1 | - | 4 | 234 | ||
| BC028528.1 | - | 4 | 252 | 1366537 | ||