siRNA Details
| Gene Symbol: NHLH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005598.3 | NP_005589.1 | 2 | 1966 | ||
| GenBank mRNA | BC013789.1 | - | 2 | 1953 | 3866861 | |
| M96739.1 | - | 2 | 1969 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NHLH1| Gene Symbol: NHLH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005598.3 | NP_005589.1 | 2 | 1966 | ||
| GenBank mRNA | BC013789.1 | - | 2 | 1953 | 3866861 | |
| M96739.1 | - | 2 | 1969 | |||