siRNA Details
| Gene Symbol: Sbsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172205.3 | NP_757342.2 | 1 | 711 | ||
| GenBank mRNA | AK081760.1 | - | 1 | 647 | ||
| AY115494.1 | - | 1 | 650 | |||
| BC139385.1 | - | 1 | 649 | 8862407 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sbsn| Gene Symbol: Sbsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172205.3 | NP_757342.2 | 1 | 711 | ||
| GenBank mRNA | AK081760.1 | - | 1 | 647 | ||
| AY115494.1 | - | 1 | 650 | |||
| BC139385.1 | - | 1 | 649 | 8862407 | ||