siRNA Details
Gene Symbol: Zfp677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172486.2 | NP_766074.2 | 4 | 2187 | ||
GenBank mRNA | AK043955.1 | - | 4 | 2189 | ||
BC057947.1 | - | 4 | 2165 | 5344769 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172486.2 | NP_766074.2 | 4 | 2187 | ||
GenBank mRNA | AK043955.1 | - | 4 | 2189 | ||
BC057947.1 | - | 4 | 2165 | 5344769 |