siRNA Details
| Gene Symbol: Dok7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172708.3 | NP_766296.1 | 6 | 1161 | ||
| GenBank mRNA | AB220919.1 | - | 7 | 1207 | ||
| AK044445.1 | - | 6 | 1162 | |||
| AK134832.1 | - | 3 | 1024 | |||
| AK170454.1 | - | 5 | 898 | |||
| BC089590.1 | - | 7 | 1280 | 6759981 | ||

Dok7
Genomic Map