siRNA Details
| Gene Symbol: Shisa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172737.4 | NP_766325.3 | 3, 4 | 1077 | ||
| GenBank mRNA | AK085127.1 | - | 2, 3 | 299 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shisa7| Gene Symbol: Shisa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172737.4 | NP_766325.3 | 3, 4 | 1077 | ||
| GenBank mRNA | AK085127.1 | - | 2, 3 | 299 | ||