siRNA Details
| Gene Symbol: Zfp647 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168276.1 | NP_001161748.1 | 6 | 1616 | ||
| NM_172817.3 | NP_766405.2 | 5 | 1531 | |||
| GenBank mRNA | AK031543.1 | - | 5 | 1532 | ||
| AY187287.1 | - | 4 | 1459 | |||
| BC055686.1 | - | 5 | 1495 | 6414212 | ||

Zfp647
Genomic Map