siRNA Details
Gene Symbol: E230025N22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172831.2 | NP_766419.2 | 8 | 894 | ||
GenBank mRNA | AK087618.1 | - | 8 | 898 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: E230025N22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172831.2 | NP_766419.2 | 8 | 894 | ||
GenBank mRNA | AK087618.1 | - | 8 | 898 |