siRNA Details
Gene Symbol: Tor1aip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172843.4 | NP_766431.3 | 5 | 1563 | ||
GenBank mRNA | AK038024.1 | - | 5 | 1825 | ||
AK080959.1 | - | 3 | 842 | |||
BC066790.1 | - | 3 | 769 | 30099444 | ||
CK635751.1 | - | 1 | 404 |
