siRNA Details
| Gene Symbol: RTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005619.4 | NP_005610.1 | 11 | 2044 | ||
| NM_206900.2 | NP_996783.1 | 10 | 1825 | |||
| NM_206901.2 | NP_996784.1 | 7 | 867 | |||
| GenBank mRNA | AF004222.1 | - | 11 | 1943 | ||
| AF004223.1 | - | 8 | 1093 | |||
| AF004224.1 | - | 7 | 835 | |||
| AF038540.1 | - | 8 | 1274 | |||
| BC073874.1 | - | 11 | 1986 | 5242827 | ||

RTN2
Genomic Map