siRNA Details
| Gene Symbol: Exd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172857.2 | NP_766445.1 | 13 | 2855 | ||
| GenBank mRNA | AK030132.1 | - | 13 | 2857 | ||
| BC092241.1 | - | 11 | 2109 | 30670567 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Exd1| Gene Symbol: Exd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172857.2 | NP_766445.1 | 13 | 2855 | ||
| GenBank mRNA | AK030132.1 | - | 13 | 2857 | ||
| BC092241.1 | - | 11 | 2109 | 30670567 | ||