siRNA Details
| Gene Symbol: Kank4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172872.3 | NP_766460.2 | 3 | 373 | ||
| GenBank mRNA | AK081525.1 | - | 3 | 311 | ||
| BC060737.1 | - | 3 | 387 | 4191502 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kank4| Gene Symbol: Kank4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172872.3 | NP_766460.2 | 3 | 373 | ||
| GenBank mRNA | AK081525.1 | - | 3 | 311 | ||
| BC060737.1 | - | 3 | 387 | 4191502 | ||