siRNA Details
| Gene Symbol: Ankk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172922.3 | NP_766510.1 | 5 | 842 | ||
| GenBank mRNA | AK036872.1 | - | 5 | 843 | ||
| BC145077.1 | - | 5 | 869 | |||
| BC145078.1 | - | 5 | 832 | 9053626 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankk1| Gene Symbol: Ankk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172922.3 | NP_766510.1 | 5 | 842 | ||
| GenBank mRNA | AK036872.1 | - | 5 | 843 | ||
| BC145077.1 | - | 5 | 869 | |||
| BC145078.1 | - | 5 | 832 | 9053626 | ||