siRNA Details
Gene Symbol: Klhl31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172925.2 | NP_766513.2 | 2 | 576 | ||
GenBank mRNA | AK053085.1 | - | 2 | 577 | ||
BC118027.1 | - | 2 | 575 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klhl31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172925.2 | NP_766513.2 | 2 | 576 | ||
GenBank mRNA | AK053085.1 | - | 2 | 577 | ||
BC118027.1 | - | 2 | 575 |