siRNA Details
| Gene Symbol: Ssc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173008.2 | NP_766596.1 | 4 | 485 | ||
| GenBank mRNA | AK079906.1 | - | 4 | 486 | ||
| AK165666.1 | - | 4 | 547 | |||
| BC141044.1 | - | 4 | 483 | 9055656 | ||
| EU850434.1 | - | 4 | 486 | |||

Ssc5d
Genomic Map