siRNA Details
| Gene Symbol: Serpina3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173024.3 | NP_766612.1 | 5 | 1309 | ||
| GenBank mRNA | AK037160.1 | - | 5 | 1311 | ||
| BC125418.1 | - | 5 | 1292 | 40129933 | ||
| BC145808.1 | - | 5 | 1292 | 40131111 | ||

Serpina3b
Genomic Map