siRNA Details
| Gene Symbol: SLCO2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005630.2 | NP_005621.2 | 14 | 3429 | ||
| XM_017007075.1 | XP_016862564.1 | 14 | 3372 | |||
| GenBank mRNA | BC041140.1 | - | 14 | 3258 | 5751516 | |
| U70867.1 | - | 14 | 3235 | |||

SLCO2A1
Genomic Map