siRNA Details
| Gene Symbol: Fam132b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173395.2 | NP_775571.2 | 8 | 1657 | ||
| GenBank mRNA | AK029264.1 | - | 8 | 1629 | ||
| BC056923.1 | - | 8 | 1657 | 5716432 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam132b| Gene Symbol: Fam132b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173395.2 | NP_775571.2 | 8 | 1657 | ||
| GenBank mRNA | AK029264.1 | - | 8 | 1629 | ||
| BC056923.1 | - | 8 | 1657 | 5716432 | ||