siRNA Details
| Gene Symbol: Sspo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173428.3 | NP_775604.3 | 9 | 1143 | ||
| GenBank mRNA | AJ491857.1 | - | 7 | 774 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sspo| Gene Symbol: Sspo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173428.3 | NP_775604.3 | 9 | 1143 | ||
| GenBank mRNA | AJ491857.1 | - | 7 | 774 | ||