siRNA Details
| Gene Symbol: Fbxo45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173439.2 | NP_775615.2 | 3 | 1362 | ||
| GenBank mRNA | BC026799.1 | - | 3 | 1434 | 3499785 | |
| BC088738.1 | - | 3 | 1362 | 6337188 | ||
| BC098203.1 | - | 3 | 1439 | 5705872 | ||

Fbxo45
Genomic Map