siRNA Details
| Gene Symbol: BC089491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175033.3 | NP_778198.2 | 1 | 236 | ||
| GenBank mRNA | AK077113.2 | - | 1 | 289 | ||
| BC089491.1 | - | 1 | 235 | 6772991 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC089491| Gene Symbol: BC089491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175033.3 | NP_778198.2 | 1 | 236 | ||
| GenBank mRNA | AK077113.2 | - | 1 | 289 | ||
| BC089491.1 | - | 1 | 235 | 6772991 | ||