siRNA Details
| Gene Symbol: Fhod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289654.1 | NP_001276583.1 | 17 | 3948 | ||
| NM_001289655.1 | NP_001276584.1 | 18 | 4305 | |||
| NM_175276.4 | NP_780485.2 | 19 | 4401 | |||
| GenBank mRNA | AB078608.1 | - | 19 | 3579 | ||
| AB078609.1 | - | 17 | 3126 | |||
| AB100088.1 | - | 18 | 3483 | |||
| AK148245.1 | - | 17 | 3090 | |||
| AK220413.1 | - | 15 | 2565 | |||
| BC056371.1 | - | 9 | 2033 | |||
| BC060607.1 | - | 4 | 1357 | |||
| BC138046.1 | - | 17 | 3048 | 8861065 | ||

Fhod3
Genomic Map