siRNA Details
| Gene Symbol: Gm4793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044378.1 | - | 2311 | |||
| BB278264.2 | - | 2 | 337 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4793| Gene Symbol: Gm4793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044378.1 | - | 2311 | |||
| BB278264.2 | - | 2 | 337 | |||