siRNA Details
| Gene Symbol: Gphb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175644.3 | NP_783575.2 | 3 | 357 | ||
| GenBank mRNA | AF467771.1 | - | 2 | 243 | ||
| AK078727.1 | - | 3 | 359 | |||
| BC111115.1 | - | 2 | 198 | |||
| BC139462.1 | - | 3 | 297 | 8862484 | ||
| BC139463.1 | - | 3 | 297 | 8862485 | ||

Gphb5
Genomic Map