siRNA Details
| Gene Symbol: Tnfrsf26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175649.5 | NP_783580.1 | 8 | 1074 | ||
| GenBank mRNA | AK142511.1 | - | 1134 | |||
| AK154666.1 | - | 8 | 1185 | |||
| AY165628.1 | - | 8 | 1074 | |||
| BC138819.1 | - | 8 | 1123 | 8861841 | ||
| BC138820.1 | - | 8 | 1124 | 8861842 | ||

Tnfrsf26
Genomic Map