siRNA Details
| Gene Symbol: Lrtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176920.4 | NP_795894.1 | 2 | 487 | ||
| GenBank mRNA | AK044492.1 | - | 2 | 441 | ||
| AK138618.1 | - | 2 | 305 | |||
| BC099587.1 | - | 2 | 285 | 30289234 | ||
| BC132594.1 | - | 2 | 191 | 40130871 | ||
| BC145196.1 | - | 2 | 191 | |||
| BC145990.1 | - | 2 | 191 | 40131293 | ||

Lrtm1
Genomic Map