siRNA Details
| Gene Symbol: EVI5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308248.1 | NP_001295177.1 | 19 | 3618 | ||
| NM_005665.5 | NP_005656.4 | 18 | 3585 | |||
| XM_017002272.1 | XP_016857761.1 | 21 | 3844 | |||
| GenBank mRNA | AB209513.1 | - | 15 | 2771 | ||
| AF008915.1 | - | 19 | 3552 | |||

EVI5
Genomic Map