siRNA Details
| Gene Symbol: Fndc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165965.1 | NP_001159437.1 | 3 | 570 | ||
| NM_177091.5 | NP_796065.2 | 3 | 570 | |||
| GenBank mRNA | AK053989.1 | - | 3 | 433 | ||
| AK054241.1 | - | 3 | 570 | |||
| BC111898.1 | - | 3 | 377 | 40061374 | ||
| BC113188.1 | - | 3 | 377 | |||

Fndc7
Genomic Map