siRNA Details
| Gene Symbol: Ccdc171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081012.1 | NP_001074481.1 | 16, 17 | 2380 | ||
| GenBank mRNA | AK039282.1 | - | 3, 4 | 498 | ||
| BC042767.1 | - | 4, 5 | 623 | |||
| BC172165.1 | - | 15, 16 | 2171 | |||

Ccdc171
Genomic Map