siRNA Details
| Gene Symbol: Iqcb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177128.4 | NP_796102.2 | 8, 9 | 950 | ||
| GenBank mRNA | AK078554.1 | - | 8, 9 | 943 | ||
| AK087791.1 | - | 1, 2 | 681 | |||
| AK165220.1 | - | 8, 9 | 934 | |||
| BC029084.1 | - | 8, 9 | 890 | 3157011 | ||

Iqcb1
Genomic Map