siRNA Details
| Gene Symbol: Fryl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036035.1 | - | 20 | 2709 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fryl| Gene Symbol: Fryl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036035.1 | - | 20 | 2709 | ||