siRNA Details
Gene Symbol: Tmem251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177140.3 | NP_796114.1 | 2 | 785 | ||
GenBank mRNA | AK032513.1 | - | 2 | 791 | ||
AK052027.1 | - | 2 | 787 | |||
AK133919.1 | - | 2 | 788 | |||
AK144149.1 | - | 788 | ||||
AK172131.1 | - | 2 | 411 | |||
BC096606.1 | - | 2 | 773 | 5366962 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_002729747.
Gene Ontology Categories:
