siRNA Details
| Gene Symbol: Rinl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177158.5 | NP_796132.2 | 10 | 1473 | ||
| GenBank mRNA | AK030993.1 | - | 10 | 1466 | ||
| AK082828.1 | - | 3 | 709 | |||
| AK162469.1 | - | 3 | 378 | |||
| AK168472.1 | - | 3 | 636 | |||
| BC044761.1 | - | 10 | 1509 | |||
| BC044878.1 | - | 1 | 190 | |||
| BC046530.1 | - | 10 | 1509 | 5329601 | ||

Rinl
Genomic Map